The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290020.1 | NP_001276949.1 | 14 - 15 | 2240 | 117 |
NM_001290021.1 | NP_001276950.1 | 7 - 8 | 1066 | 117 | |
NM_001312689.1 | NP_001299618.1 | 9 - 10 | 1457 | 117 | |
NM_018076.4 | NP_060546.2 | 14 - 15 | 2193 | 117 | |
XM_011519527.1 | XP_011517829.1 | 14 - 15 | 2240 | 117 | |
XM_011519528.1 | XP_011517830.1 | 14 - 15 | 2890 | 117 | |
XM_011519533.2 | XP_011517835.1 | 9 - 10 | 1414 | 117 | |
XM_011519534.2 | XP_011517836.1 | 9 - 10 | 1547 | 117 | |
XM_011519535.2 | XP_011517837.1 | 8 - 9 | 1299 | 117 | |
XM_011519537.2 | XP_011517839.1 | 5 - 6 | 974 | 117 | |
XM_017016368.1 | XP_016871857.1 | 14 - 15 | 2240 | 117 | |
XM_017016369.1 | XP_016871858.1 | 14 - 15 | 2240 | 117 | |
XM_017016370.1 | XP_016871859.1 | 13 - 14 | 2030 | 117 | |
GenBank mRNA | AK001679.1 | - | 14 - 15 | 2193 | 117 |
AK292521.1 | - | 14 - 15 | 2196 | 117 | |
AK302048.1 | - | 7 - 8 | 1020 | 117 | |
AK302419.1 | - | 9 - 10 | 1789 | 117 | |
AK302627.1 | - | 9 - 10 | 1457 | 117 | |
AL136859.1 | - | 3 - 4 | 579 | 117 | |
BC140846.1 | - | 13 - 14 | 2121 | 117 |