Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FLYWCH1
Entrez Gene ID
84256
Gene Name
FLYWCH-type zinc finger 1
Gene Aliases
-
Chromosome Location
Chr.16: 2911873 - 2951208 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308068.1 NP_001294997.1 9 - 10 2476 75
NM_032296.2 NP_115672.2 9 - 10 2473 75
XM_006720962.3 XP_006721025.1 11 - 12 2725 75
XM_011522701.2 XP_011521003.1 10 - 11 3114 75
XM_011522702.2 XP_011521004.1 10 - 11 3588 75
XM_011522703.2 XP_011521005.1 11 - 12 3209 75
XM_011522704.2 XP_011521006.1 10 - 11 3112 75
XM_011522705.2 XP_011521007.1 11 - 12 2870 75
XM_017023772.1 XP_016879261.1 12 - 13 3306 75
GenBank mRNA AB046772.1 - 9 - 10 2453 75
AK026885.1 - 4 - 5 642 75
AK055777.1 - 3 - 4 1238 75
AK057022.1 - 4 - 5 1953 75
AK092434.1 - 5 - 6 1293 75
AK127235.1 - 4 - 5 2597 75
AL136585.1 - 7 - 8 1445 75
BC018712.1 - 5 - 6 1221 75
BC028572.1 - 9 - 10 2477 75

View Full Product Details