The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304353.1 | NP_001291282.1 | 3 - 4 | 358 | 76 |
NM_001304354.1 | NP_001291283.1 | 3 - 4 | 355 | 76 | |
NM_032309.3 | NP_115685.1 | 3 - 4 | 521 | 76 | |
GenBank mRNA | BC004498.2 | - | 3 - 4 | 499 | 76 |
BM563093.1 | - | 3 - 4 | 355 | 76 | |
BX115079.1 | - | 3 - 4 | 357 | 76 | |
N39730.1 | - | 3 - 4 | 355 | 76 |