Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MEGF10
Entrez Gene ID
84466
Gene Name
multiple EGF like domains 10
Gene Aliases
EMARDD
Chromosome Location
Chr.5: 127229519 - 127461222 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256545.1 NP_001243474.1 2 - 3 432 82
NM_001308119.1 NP_001295048.1 3 - 4 483 82
NM_001308121.1 NP_001295050.1 2 - 3 432 82
NM_032446.2 NP_115822.1 3 - 4 483 82
XM_011543694.1 XP_011541996.1 2 - 3 195 82
XM_017009987.1 XP_016865476.1 4 - 5 728 82
GenBank mRNA AB058676.1 - 3 - 4 324 82
AK123568.1 - 4 - 5 598 82
AK307919.1 - 2 - 3 351 82
BC020198.1 - 3 - 4 400 82
BC152478.1 - 3 - 4 324 82
CR749437.1 - 2 - 3 366 82
DA240513.1 - 2 - 3 432 82

View Full Product Details