The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000738.2 | NP_000729.2 | 2 - 2 | 1095 | 82 |
XM_011544742.2 | XP_011543044.1 | 2 - 2 | 796 | 82 | |
GenBank mRNA | AF385587.1 | - | 1 - 1 | 704 | 82 |
AF498915.1 | - | 1 - 1 | 694 | 82 | |
AK312374.1 | - | 2 - 2 | 856 | 82 | |
BC007740.2 | - | 2 - 2 | 816 | 82 | |
BC022984.1 | - | 2 - 2 | 1095 | 82 | |
BT007166.1 | - | 1 - 1 | 694 | 82 |