The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001692.3 | NP_001683.2 | 8 - 9 | 877 | 90 |
XM_011532907.2 | XP_011531209.1 | 7 - 8 | 1089 | 90 | |
GenBank mRNA | AK127853.1 | - | 10 - 11 | 2617 | 90 |
AK223151.1 | - | 8 - 9 | 821 | 90 | |
AK291121.1 | - | 8 - 9 | 886 | 90 | |
AK313194.1 | - | 8 - 9 | 853 | 90 | |
BC063411.1 | - | 8 - 9 | 854 | 90 | |
M25809.1 | - | 8 - 9 | 839 | 90 |