The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002565.3 | NP_002556.1 | 1 - 1 | 729 | 91 |
GenBank mRNA | BC095503.1 | - | 1 - 1 | 688 | 91 |
BC096067.3 | - | 1 - 1 | 592 | 91 | |
BC096068.1 | - | 1 - 1 | 592 | 91 | |
BC096069.1 | - | 1 - 1 | 592 | 91 | |
BC096070.1 | - | 1 - 1 | 592 | 91 | |
FJ599701.1 | - | 1 - 1 | 298 | 91 | |
FJ599702.1 | - | 1 - 1 | 294 | 91 |