The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003528.2 | NP_003519.1 | 1 - 1 | 835 | 55 |
GenBank mRNA | AK093747.1 | - | 2 - 2 | 795 | 55 |
AK289725.1 | - | 2 - 2 | 1193 | 55 | |
BC005827.2 | - | 1 - 1 | 805 | 55 | |
BC069193.1 | - | 1 - 1 | 817 | 55 | |
M60756.1 | - | 1 - 1 | 716 | 55 |