Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VPS45
Entrez Gene ID
11311
Gene Name
vacuolar protein sorting 45 homolog
Gene Aliases
H1, H1VPS45, SCN5, VPS45A, VPS45B, VPS54A, VSP45, VSP45A
Chromosome Location
Chr.1: 150067293 - 150145327 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001279353.1 NP_001266282.1 7 - 8 952 91
NM_001279354.1 NP_001266283.1 9 - 10 1148 91
NM_001279355.1 NP_001266284.1 6 - 7 759 91
NM_007259.4 NP_009190.2 9 - 10 1503 91
NR_103998.1 7 - 8 921 91
XM_017000164.1 XP_016855653.1 9 - 10 1053 91
GenBank mRNA AF165513.1 - 9 - 10 983 91
AJ133421.1 - 9 - 10 971 91
AK023170.1 - 9 - 10 990 91
AK223214.1 - 9 - 10 956 91
AK293555.1 - 7 - 8 782 91
AK295529.1 - 9 - 10 91
AK298786.1 - 7 - 8 863 91
AK298835.1 - 6 - 7 619 91
AY453586.1 - 3 - 4 842 91
BC012932.1 - 9 - 10 989 91
BC028382.2 - 9 - 10 1484 91

View Full Product Details