The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276342.1 | NP_001263271.1 | 8 - 9 | 965 | 128 |
NM_001318707.1 | NP_001305636.1 | 7 - 8 | 880 | 128 | |
NM_001318708.1 | NP_001305637.1 | 9 - 10 | 1024 | 128 | |
NM_001318709.1 | NP_001305638.1 | 8 - 9 | 965 | 128 | |
NM_013326.4 | NP_037458.3 | 10 - 11 | 1109 | 128 | |
XM_011525939.1 | XP_011524241.1 | 9 - 10 | 981 | 128 | |
XM_017025717.1 | XP_016881206.1 | 8 - 9 | 918 | 128 | |
GenBank mRNA | AF143536.1 | - | 9 - 10 | 856 | 128 |
AK027106.1 | - | 2 - 3 | 184 | 128 | |
AK057192.1 | - | 10 - 11 | 1070 | 128 | |
AK294169.1 | - | 7 - 8 | 827 | 128 | |
AK295246.1 | - | 9 - 10 | 981 | 128 | |
AK296230.1 | - | 8 - 9 | 916 | 128 | |
BC002950.1 | - | 10 - 11 | 1034 | 128 | |
BC008305.1 | - | 10 - 11 | 1041 | 128 |