Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCL6
Entrez Gene ID
604
Gene Name
B-cell CLL/lymphoma 6
Gene Aliases
BCL5, BCL6A, LAZ3, ZBTB27, ZNF51
Chromosome Location
Chr.3: 187721377 - 187745725 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130845.1 NP_001124317.1 9 - 10 2434 63
NM_001134738.1 NP_001128210.1 7 - 8 1858 63
NM_001706.4 NP_001697.2 9 - 10 2342 63
XM_005247694.3 XP_005247751.1 9 - 10 5686 63
XM_011513062.2 XP_011511364.1 8 - 9 2174 63
GenBank mRNA AK304325.1 - 9 - 10 2001 63
AK308162.1 - 9 - 10 2083 63
BC142705.1 - 9 - 10 2057 63
BC150184.1 - 9 - 10 2058 63
BX649185.1 - 9 - 10 2964 63
EU139066.1 - 6 - 7 1833 63
EU883531.1 - 6 - 7 1809 63
S67779.1 - 9 - 10 2396 63
U00115.1 - 9 - 10 2304 63
Z21943.1 - 9 - 10 2061 63

View Full Product Details