The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173495.2 | NP_775766.2 | 2 - 3 | 1015 | 108 |
XM_011545449.2 | XP_011543751.1 | 3 - 4 | 1100 | 108 | |
GenBank mRNA | AK054858.1 | - | 2 - 3 | 905 | 108 |
AK298796.1 | - | 2 - 3 | 1018 | 108 | |
BC121061.2 | - | 2 - 3 | 862 | 108 | |
KJ534921.1 | - | 2 - 3 | 1007 | 108 | |
KJ535084.1 | - | 3 - 4 | 966 | 108 | |
KJ535090.1 | - | 3 - 4 | 1171 | 108 |