Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC7A13
Entrez Gene ID
157724
Gene Name
solute carrier family 7 member 13
Gene Aliases
AGT-1, AGT1, XAT2
Chromosome Location
Chr.8: 86213909 - 86230380 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138817.2 NP_620172.2 1 - 2 784 88
GenBank mRNA AJ417661.1 - 1 - 2 689 88
AK055815.1 - 1 - 2 784 88
BC125165.1 - 1 - 2 769 88

View Full Product Details