The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031215.2 | NP_112492.2 | 3 - 4 | 534 | 76 |
GenBank mRNA | BC003122.1 | - | 1 - 2 | 69 | 76 |
BU166894.1 | - | 3 - 4 | 247 | 76 | |
CB960872.1 | - | 3 - 4 | 564 | 76 |