The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138370.2 | NP_612379.2 | 5 - 6 | 1399 | 81 |
GenBank mRNA | BC007901.2 | - | 3 - 4 | 354 | 81 |
BC062988.1 | - | 3 - 4 | 354 | 81 | |
BC094697.1 | - | 5 - 6 | 807 | 81 | |
BC110513.1 | - | 5 - 6 | 836 | 81 | |
BC110514.1 | - | 5 - 6 | 836 | 81 |