The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146726.1 | NP_001140198.1 | 2 - 3 | 460 | 65 |
NM_138379.2 | NP_612388.2 | 2 - 3 | 460 | 65 | |
XM_011534694.2 | XP_011532996.1 | 2 - 3 | 431 | 65 | |
XM_017010021.1 | XP_016865510.1 | 2 - 3 | 431 | 65 | |
GenBank mRNA | AK301337.1 | - | 2 - 3 | 431 | 65 |
AK312746.1 | - | 2 - 3 | 430 | 65 | |
BC008988.1 | - | 2 - 3 | 460 | 65 | |
JX049980.1 | - | 2 - 3 | 403 | 65 | |
KU178832.1 | - | 2 - 3 | 403 | 65 | |
KU178833.1 | - | 2 - 3 | 403 | 65 |