Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IZUMO4
Entrez Gene ID
113177
Gene Name
IZUMO family member 4
Gene Aliases
C19orf36, IMAGE:4215339
Chromosome Location
Chr.19: 2096848 - 2099590 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031735.2 NP_001026905.2 3 - 4 450 99
NM_001039846.1 NP_001034935.1 3 - 4 450 99
XM_005259480.3 XP_005259537.2 3 - 4 471 99
GenBank mRNA AK294083.1 - 3 - 4 321 99
AK301957.1 - 3 - 4 404 195
AW182303.1 - 5 - 6 99
AY753639.1 - 3 - 4 409 99
BC010443.1 - 3 - 4 389 99
BC014609.2 - 3 - 4 404 99
BC040964.1 - 3 - 4 391 99
BI827974.1 - 3 - 4 306 99
BX378854.2 - 3 - 4 450 99
DQ196309.1 - 3 - 4 437 99
DQ196310.1 - 3 - 4 437 99
DQ424900.1 - 3 - 4 373 99

View Full Product Details