Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CASC4
Entrez Gene ID
113201
Gene Name
cancer susceptibility candidate 4
Gene Aliases
H63
Chromosome Location
Chr.15: 44288616 - 44415761 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC012124.1 - 3 - 4 788 116

View Full Product Details