Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM98A
Entrez Gene ID
25940
Gene Name
family with sequence similarity 98 member A
Gene Aliases
-
Chromosome Location
Chr.2: 33583661 - 33599362 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001304538.1 NP_001291467.1 5 - 6 679 62
NM_015475.4 NP_056290.3 6 - 7 864 62
GenBank mRNA AK074867.1 - 6 - 7 800 62
AK300023.1 - 5 - 6 608 62
AK302233.1 - 2 - 3 287 62
AL049943.1 - 1 - 2 111 62
BC038581.1 - 3 - 4 397 62
BC060860.1 - 6 - 7 777 62
BC136768.1 - 6 - 7 768 62
BC136770.1 - 6 - 7 768 62
CR936766.1 - 6 - 7 802 62

View Full Product Details