Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PTPN20
- Entrez Gene ID
-
26095
- Gene Name
- protein tyrosine phosphatase, non-receptor type 20
- Gene Aliases
-
CT126, PTPN20A, PTPN20B, bA142I17.1, bA42B19.1
- Chromosome Location
-
Chr.10: 46911134 - 47003834 on Build GRCh38
View Full Product Details