The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015431.3 | NP_056246.3 | 3 - 4 | 794 | 104 |
GenBank mRNA | AF327057.1 | - | 3 - 4 | 794 | 104 |
AK096188.1 | - | 3 - 4 | 676 | 104 | |
BC074748.2 | - | 2 - 3 | 338 | 104 | |
BX644877.1 | - | 3 - 4 | 350 | 104 |