Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SORL1
Entrez Gene ID
6653
Gene Name
sortilin related receptor 1
Gene Aliases
C11orf32, LR11, LRP9, SORLA, SorLA-1, gp250
Chromosome Location
Chr.11: 121452203 - 121633762 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003105.5 NP_003096.1 48 - 48 9765 60
XM_011542963.2 XP_011541265.1 47 - 47 9526 60
XM_011542965.2 XP_011541267.1 42 - 42 8674 60
XM_011542967.2 XP_011541269.1 28 - 28 6706 60
XM_017018169.1 XP_016873658.1 48 - 48 9472 60
XM_017018170.1 XP_016873659.1 46 - 46 9148 60
XM_017018172.1 XP_016873661.1 34 - 34 7637 60
GenBank mRNA AK025583.1 - 1 - 1 1496 60
AK096577.1 - 1 - 1 2571 60
BC040643.1 - 1 - 1 3067 60
U90916.1 - 1 - 1 750 60

View Full Product Details