The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012335.3 | NP_036467.2 | 14 - 15 | 1666 | 70 |
XM_011528024.1 | XP_011526326.1 | 14 - 15 | 1666 | 70 | |
XM_011528025.1 | XP_011526327.1 | 14 - 15 | 1654 | 70 | |
XM_011528026.1 | XP_011526328.1 | 13 - 14 | 1579 | 70 | |
XM_017026821.1 | XP_016882310.1 | 14 - 15 | 1654 | 70 | |
GenBank mRNA | AB210037.1 | - | 14 - 15 | 1665 | 70 |
AB290178.1 | - | 14 - 15 | 1796 | 70 | |
AJ310570.1 | - | 14 - 15 | 1568 | 70 | |
AK131106.1 | - | 14 - 15 | 1665 | 70 | |
BC028071.1 | - | 14 - 15 | 1642 | 70 | |
X98411.1 | - | 5 - 6 | 577 | 70 |