Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 236482175-236482678 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDARADD
Entrez Gene ID
128178
Gene Name
EDAR associated death domain
Gene Aliases
ECTD11A, ECTD11B, ED3, EDA3
UniGene
Hs.352224
Gene Chromosome Location
Chr.1: 236394380-236484708 on Build GRCh38
Transcripts
NM_080738.3 NM_145861.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
116919 c.382A>C p.K128Q Chr.1: 236482383-236482383 on Build GRCh38
6062121 c.636C>T p.S212S Chr.1: 236482637-236482637 on Build GRCh38
373956 c.513C>A p.N171K Chr.1: 236482514-236482514 on Build GRCh38
6062125 c.328C>A p.P110T Chr.1: 236482329-236482329 on Build GRCh38
5845935 c.617G>A p.R206Q Chr.1: 236482618-236482618 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs900227433 Chr.1: 236482655-236482655 on Build GRCh38
rs780289873 Chr.1: 236482253-236482253 on Build GRCh38
rs750157040 Chr.1: 236482623-236482623 on Build GRCh38
rs936740826 Chr.1: 236482338-236482338 on Build GRCh38
rs769122607 Chr.1: 236482405-236482405 on Build GRCh38
rs376972393 Chr.1: 236482379-236482379 on Build GRCh38
rs201032777 Chr.1: 236482462-236482462 on Build GRCh38
rs954823206 Chr.1: 236482418-236482418 on Build GRCh38
rs761026610 Chr.1: 236482259-236482259 on Build GRCh38
rs147501905 Chr.1: 236482371-236482371 on Build GRCh38
rs757216585 Chr.1: 236482617-236482617 on Build GRCh38
rs755197423 Chr.1: 236482634-236482634 on Build GRCh38
rs779277524 Chr.1: 236482376-236482376 on Build GRCh38
rs777899133 Chr.1: 236482356-236482356 on Build GRCh38
rs777298254 Chr.1: 236482456-236482456 on Build GRCh38
rs147249573 Chr.1: 236482323-236482323 on Build GRCh38
rs768783242 Chr.1: 236482562-236482562 on Build GRCh38
rs755966826 Chr.1: 236482350-236482350 on Build GRCh38
rs759679168 Chr.1: 236482244-236482244 on Build GRCh38
rs74942492 Chr.1: 236482601-236482601 on Build GRCh38
rs906737114 Chr.1: 236482349-236482349 on Build GRCh38
rs1030214435 Chr.1: 236482414-236482414 on Build GRCh38
rs374279273 Chr.1: 236482604-236482604 on Build GRCh38
rs762203601 Chr.1: 236482295-236482295 on Build GRCh38
rs763626415 Chr.1: 236482602-236482602 on Build GRCh38
rs764680952 Chr.1: 236482261-236482261 on Build GRCh38
rs371337563 Chr.1: 236482493-236482493 on Build GRCh38
rs747038830 Chr.1: 236482552-236482552 on Build GRCh38
rs483352767 Chr.1: 236482588-236482588 on Build GRCh38
rs969366797 Chr.1: 236482311-236482311 on Build GRCh38
rs765992938 Chr.1: 236482299-236482299 on Build GRCh38
rs767229616 Chr.1: 236482466-236482466 on Build GRCh38
rs772349212 Chr.1: 236482392-236482392 on Build GRCh38
rs771250194 Chr.1: 236482233-236482233 on Build GRCh38
rs758511689 Chr.1: 236482528-236482528 on Build GRCh38
rs753890063 Chr.1: 236482494-236482494 on Build GRCh38
rs780366105 Chr.1: 236482532-236482532 on Build GRCh38
rs121908116 Chr.1: 236482366-236482366 on Build GRCh38
rs371739998 Chr.1: 236482227-236482227 on Build GRCh38
rs749128914 Chr.1: 236482228-236482228 on Build GRCh38
rs777172467 Chr.1: 236482406-236482406 on Build GRCh38
rs144235068 Chr.1: 236482648-236482648 on Build GRCh38
rs773408614 Chr.1: 236482579-236482579 on Build GRCh38
rs397515575 Chr.1: 236482403-236482408 on Build GRCh38
rs373971442 Chr.1: 236482283-236482283 on Build GRCh38
rs114632254 Chr.1: 236482309-236482309 on Build GRCh38
rs752275345 Chr.1: 236482324-236482324 on Build GRCh38
rs604070 Chr.1: 236482370-236482370 on Build GRCh38
rs373028114 Chr.1: 236482568-236482568 on Build GRCh38
rs760843174 Chr.1: 236482400-236482400 on Build GRCh38
rs369166930 Chr.1: 236482245-236482245 on Build GRCh38
rs371177909 Chr.1: 236482566-236482566 on Build GRCh38
rs758607876 Chr.1: 236482399-236482403 on Build GRCh38
rs544038801 Chr.1: 236482226-236482226 on Build GRCh38
rs780323827 Chr.1: 236482643-236482643 on Build GRCh38
rs377531010 Chr.1: 236482526-236482526 on Build GRCh38
rs139996586 Chr.1: 236482394-236482394 on Build GRCh38
rs755696159 Chr.1: 236482478-236482478 on Build GRCh38
rs376675770 Chr.1: 236482239-236482239 on Build GRCh38
rs74315309 Chr.1: 236482455-236482455 on Build GRCh38
rs781199161 Chr.1: 236482647-236482647 on Build GRCh38
rs1005957166 Chr.1: 236482393-236482393 on Build GRCh38
rs754505568 Chr.1: 236482322-236482322 on Build GRCh38
rs928145231 Chr.1: 236482581-236482581 on Build GRCh38
rs534068767 Chr.1: 236482279-236482279 on Build GRCh38
rs1031441787 Chr.1: 236482450-236482450 on Build GRCh38
rs753408117 Chr.1: 236482616-236482616 on Build GRCh38
rs752115447 Chr.1: 236482469-236482469 on Build GRCh38
rs763414523 Chr.1: 236482587-236482587 on Build GRCh38
rs765300638 Chr.1: 236482618-236482618 on Build GRCh38
rs886046186 Chr.1: 236482303-236482303 on Build GRCh38
rs143891636 Chr.1: 236482458-236482458 on Build GRCh38
rs200017138 Chr.1: 236482571-236482571 on Build GRCh38
rs375789023 Chr.1: 236482572-236482572 on Build GRCh38
rs747324152 Chr.1: 236482633-236482633 on Build GRCh38
rs199965336 Chr.1: 236482430-236482430 on Build GRCh38
rs747968826 Chr.1: 236482220-236482220 on Build GRCh38
rs757261515 Chr.1: 236482510-236482510 on Build GRCh38
rs797045087 Chr.1: 236482300-236482301 on Build GRCh38
rs750512955 Chr.1: 236482267-236482268 on Build GRCh38
rs995250116 Chr.1: 236482556-236482556 on Build GRCh38
rs773579559 Chr.1: 236482454-236482454 on Build GRCh38
rs373326410 Chr.1: 236482487-236482487 on Build GRCh38
rs778797256 Chr.1: 236482522-236482522 on Build GRCh38
rs879255629 Chr.1: 236482368-236482368 on Build GRCh38
rs748587639 Chr.1: 236482569-236482569 on Build GRCh38
rs369614195 Chr.1: 236482451-236482451 on Build GRCh38
rs376731607 Chr.1: 236482624-236482624 on Build GRCh38

View Full Product Details