Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 236482568-236483070 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDARADD
Entrez Gene ID
128178
Gene Name
EDAR associated death domain
Gene Aliases
ECTD11A, ECTD11B, ED3, EDA3
UniGene
Hs.352224
Gene Chromosome Location
Chr.1: 236394380-236484708 on Build GRCh38
Transcripts
NM_080738.3 NM_145861.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6062121 c.636C>T p.S212S Chr.1: 236482637-236482637 on Build GRCh38
5845935 c.617G>A p.R206Q Chr.1: 236482618-236482618 on Build GRCh38
6024915 c.628G>T p.D210Y Chr.1: 236482629-236482629 on Build GRCh38
5784164 c.576G>T p.E192D Chr.1: 236482607-236482607 on Build GRCh38
3485612 c.576G>A p.E192E Chr.1: 236482577-236482577 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774204933 Chr.1: 236482750-236482750 on Build GRCh38
rs761943296 Chr.1: 236483007-236483007 on Build GRCh38
rs900227433 Chr.1: 236482655-236482655 on Build GRCh38
rs928152450 Chr.1: 236482914-236482914 on Build GRCh38
rs983460041 Chr.1: 236482932-236482932 on Build GRCh38
rs750157040 Chr.1: 236482623-236482623 on Build GRCh38
rs886046189 Chr.1: 236483000-236483001 on Build GRCh38
rs1039802466 Chr.1: 236482792-236482792 on Build GRCh38
rs757623127 Chr.1: 236482738-236482738 on Build GRCh38
rs1046804913 Chr.1: 236482815-236482815 on Build GRCh38
rs771390820 Chr.1: 236482667-236482667 on Build GRCh38
rs775159272 Chr.1: 236482844-236482844 on Build GRCh38
rs774580843 Chr.1: 236482670-236482670 on Build GRCh38
rs568663942 Chr.1: 236482661-236482661 on Build GRCh38
rs757216585 Chr.1: 236482617-236482617 on Build GRCh38
rs755197423 Chr.1: 236482634-236482634 on Build GRCh38
rs941708172 Chr.1: 236482939-236482939 on Build GRCh38
rs766538054 Chr.1: 236482768-236482768 on Build GRCh38
rs74942492 Chr.1: 236482601-236482601 on Build GRCh38
rs539403998 Chr.1: 236482927-236482927 on Build GRCh38
rs1024732136 Chr.1: 236482920-236482920 on Build GRCh38
rs771924946 Chr.1: 236482765-236482765 on Build GRCh38
rs1023731788 Chr.1: 236482783-236482783 on Build GRCh38
rs374279273 Chr.1: 236482604-236482604 on Build GRCh38
rs760609411 Chr.1: 236482921-236482921 on Build GRCh38
rs999877223 Chr.1: 236482749-236482749 on Build GRCh38
rs763626415 Chr.1: 236482602-236482602 on Build GRCh38
rs79578216 Chr.1: 236482778-236482778 on Build GRCh38
rs933479840 Chr.1: 236482724-236482724 on Build GRCh38
rs483352767 Chr.1: 236482588-236482588 on Build GRCh38
rs770253410 Chr.1: 236482658-236482658 on Build GRCh38
rs765675921 Chr.1: 236482774-236482774 on Build GRCh38
rs142266831 Chr.1: 236482934-236482934 on Build GRCh38
rs554006482 Chr.1: 236482837-236482837 on Build GRCh38
rs910820776 Chr.1: 236482756-236482756 on Build GRCh38
rs572424730 Chr.1: 236482841-236482841 on Build GRCh38
rs886046187 Chr.1: 236482749-236482749 on Build GRCh38
rs768449995 Chr.1: 236482679-236482679 on Build GRCh38
rs898160289 Chr.1: 236482802-236482802 on Build GRCh38
rs144235068 Chr.1: 236482648-236482648 on Build GRCh38
rs576451942 Chr.1: 236482955-236482955 on Build GRCh38
rs1021069081 Chr.1: 236482975-236482975 on Build GRCh38
rs1051910969 Chr.1: 236482746-236482746 on Build GRCh38
rs776244509 Chr.1: 236482680-236482680 on Build GRCh38
rs1037364729 Chr.1: 236482971-236482971 on Build GRCh38
rs1052073187 Chr.1: 236482662-236482662 on Build GRCh38
rs1013389851 Chr.1: 236482851-236482851 on Build GRCh38
rs780323827 Chr.1: 236482643-236482643 on Build GRCh38
rs781199161 Chr.1: 236482647-236482647 on Build GRCh38
rs761452687 Chr.1: 236482684-236482684 on Build GRCh38
rs557974993 Chr.1: 236482969-236482970 on Build GRCh38
rs753408117 Chr.1: 236482616-236482616 on Build GRCh38
rs765300638 Chr.1: 236482618-236482618 on Build GRCh38
rs747324152 Chr.1: 236482633-236482633 on Build GRCh38
rs886046190 Chr.1: 236483003-236483003 on Build GRCh38
rs749600700 Chr.1: 236482663-236482663 on Build GRCh38
rs753919866 Chr.1: 236482998-236482998 on Build GRCh38
rs931091068 Chr.1: 236482812-236482812 on Build GRCh38
rs886046188 Chr.1: 236482764-236482764 on Build GRCh38
rs759399160 Chr.1: 236482752-236482752 on Build GRCh38
rs969493258 Chr.1: 236482810-236482810 on Build GRCh38
rs530109023 Chr.1: 236482811-236482811 on Build GRCh38
rs760342049 Chr.1: 236482673-236482673 on Build GRCh38
rs765066608 Chr.1: 236482685-236482685 on Build GRCh38
rs376731607 Chr.1: 236482624-236482624 on Build GRCh38

View Full Product Details