The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014010.4 | NP_054729.3 | 1 - 2 | 544 | 66 |
GenBank mRNA | AB014534.1 | - | 1 - 2 | 544 | 66 |
BC146756.1 | - | 1 - 2 | 544 | 66 |