The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020700.1 | NP_065751.1 | 3 - 4 | 909 | 85 |
XM_011538578.2 | XP_011536880.1 | 3 - 4 | 1045 | 85 | |
XM_011538579.1 | XP_011536881.1 | 3 - 4 | 597 | 85 | |
XM_017019676.1 | XP_016875165.1 | 3 - 4 | 1159 | 85 | |
GenBank mRNA | AB032983.1 | - | 2 - 3 | 502 | 85 |
AB084258.1 | - | 3 - 4 | 1174 | 85 | |
AK023315.1 | - | 4 - 5 | 971 | 85 | |
BC157843.1 | - | 3 - 4 | 919 | 85 |