The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042368.2 | NP_001035827.1 | 9 - 10 | 1682 | 107 |
NM_001271774.1 | NP_001258703.1 | 9 - 10 | 1594 | 107 | |
NM_001271775.1 | NP_001258704.1 | 9 - 10 | 1687 | 107 | |
NM_001271776.1 | NP_001258705.1 | 9 - 10 | 1667 | 107 | |
NM_006266.3 | NP_006257.1 | 9 - 10 | 1703 | 107 | |
GenBank mRNA | AB037729.1 | - | 5 - 6 | 995 | 107 |
AF027169.1 | - | 9 - 10 | 1598 | 107 | |
AF295773.1 | - | 9 - 10 | 1618 | 107 | |
AK000242.1 | - | 4 - 5 | 660 | 107 | |
AK024575.1 | - | 1 - 2 | 257 | 107 | |
AK056462.1 | - | 8 - 9 | 1380 | 107 | |
AK074114.1 | - | 9 - 10 | 1573 | 107 | |
AK090450.1 | - | 5 - 6 | 1029 | 107 | |
AK127524.1 | - | 9 - 10 | 1687 | 107 | |
AK301470.1 | - | 9 - 10 | 1554 | 107 | |
BC033198.1 | - | 1 - 2 | 99 | 107 | |
BC059362.1 | - | 9 - 10 | 1657 | 107 |