The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178123.4 | NP_835224.3 | 18 - 18 | 9205 | 69 |
XM_011512139.2 | XP_011510441.1 | 18 - 18 | 9233 | 69 | |
XM_011512141.2 | XP_011510443.1 | 16 - 16 | 8910 | 69 | |
XM_017005250.1 | XP_016860739.1 | 17 - 17 | 9059 | 69 | |
XM_017005251.1 | XP_016860740.1 | 19 - 19 | 9411 | 69 | |
GenBank mRNA | AK056075.1 | - | 1 - 1 | 1550 | 69 |