Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DA699339.1 - 1 353 Chr. 12 - 48189349 - 48189874 [+] on Build GRCh38 70
GenBankmRNA AK056127.1 - 1 353 Chr. 12 - 48189349 - 48191207 [+] on Build GRCh38 70


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream coiled-coil domain containing 184 CCDC184 C12orf68 -3.4kb Sense Chr. - 48183583 - 48185926 on Build GRCh38
Downstream olfactory receptor family 10 subfamily AD member 1 OR10AD1 OR10AD1P, OR12-1 10.6kb Anti-sense Chr. - 48202339 - 48203292 on Build GRCh38

View Full Product Details