Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNRNP48
Entrez Gene ID
154007
Gene Name
small nuclear ribonucleoprotein U11/U12 subunit 48
Gene Aliases
C6orf151, dJ336K20B.1, dJ512B11.2
Chromosome Location
Chr.6: 7590199 - 7611967 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152551.3 NP_689764.3 1 - 2 215 79
GenBank mRNA AK056796.1 - 1 - 2 192 79
AK292409.1 - 1 - 2 193 79
AL832462.1 - 1 - 2 216 79
BC054008.1 - 1 - 2 188 79
BC062458.1 - 1 - 2 177 79
BC114630.1 - 1 - 2 183 79
BC114631.1 - 1 - 2 184 79

View Full Product Details