The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144650.2 | NP_653251.2 | 8 - 9 | 768 | 68 |
GenBank mRNA | AK056992.1 | - | 7 - 8 | 705 | 68 |
AK293530.1 | - | 5 - 6 | 608 | 68 | |
AK294114.1 | - | 6 - 7 | 471 | 68 | |
AK300243.1 | - | 8 - 9 | 747 | 68 | |
AK300881.1 | - | 7 - 8 | 554 | 68 | |
AY033237.1 | - | 8 - 9 | 753 | 68 | |
BG698892.1 | - | 3 - 4 | 310 | 68 |