The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204344.1 | NP_001191273.1 | 2 - 2 | 183 | 97 |
NM_005118.3 | NP_005109.2 | 4 - 4 | 476 | 97 | |
GenBank mRNA | AF039390.1 | - | 1 - 1 | 1252 | 97 |
AF520785.1 | - | 4 - 4 | 482 | 97 | |
AK291642.1 | - | 4 - 4 | 387 | 97 | |
AY434464.1 | - | 2 - 2 | 183 | 97 | |
BC069435.1 | - | 4 - 4 | 431 | 97 | |
BC074940.2 | - | 4 - 4 | 400 | 97 | |
BC074941.2 | - | 4 - 4 | 400 | 97 | |
BC104462.1 | - | 4 - 4 | 475 | 97 | |
BC104463.1 | - | 4 - 4 | 476 | 97 |