The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000442.4 | NP_000433.4 | 1 - 2 | 277 | 142 |
XM_005276880.1 | XP_005276937.1 | 1 - 2 | 563 | 142 | |
XM_005276881.1 | XP_005276938.1 | 1 - 2 | 563 | 142 | |
XM_005276882.1 | XP_005276939.1 | 1 - 2 | 563 | 142 | |
XM_005276883.2 | XP_005276940.1 | 1 - 2 | 563 | 142 | |
XM_011524889.2 | XP_011523191.1 | 2 - 3 | 293 | 142 | |
XM_011524890.1 | XP_011523192.1 | 2 - 3 | 242 | 142 | |
XM_017024738.1 | XP_016880227.1 | 2 - 3 | 186 | 142 | |
XM_017024739.1 | XP_016880228.1 | 1 - 2 | 563 | 142 | |
XM_017024740.1 | XP_016880229.1 | 2 - 3 | 256 | 142 | |
XM_017024741.1 | XP_016880230.1 | 1 - 2 | 563 | 142 | |
GenBank mRNA | AF281287.1 | - | 1 - 2 | 147 | 142 |
AF281288.1 | - | 1 - 2 | 147 | 142 | |
AF281289.1 | - | 1 - 2 | 147 | 142 | |
AF281290.1 | - | 1 - 2 | 147 | 142 | |
AF281291.1 | - | 1 - 2 | 147 | 142 | |
AF281292.1 | - | 1 - 2 | 147 | 142 | |
AF281293.1 | - | 1 - 2 | 147 | 142 | |
AF281294.1 | - | 1 - 2 | 147 | 142 | |
AF281295.1 | - | 1 - 2 | 147 | 142 | |
AF281296.1 | - | 1 - 2 | 147 | 142 | |
AF281297.1 | - | 1 - 2 | 147 | 142 | |
AF281298.1 | - | 1 - 2 | 147 | 142 | |
AF281299.1 | - | 1 - 2 | 147 | 142 | |
AF281300.1 | - | 1 - 2 | 147 | 142 | |
AF281301.1 | - | 1 - 2 | 147 | 142 | |
AF393676.1 | - | 1 - 2 | 147 | 142 | |
AF393677.1 | - | 1 - 2 | 147 | 142 | |
AF393678.1 | - | 1 - 2 | 147 | 142 | |
BC022512.1 | - | 1 - 2 | 257 | 142 | |
BC051822.1 | - | 1 - 2 | 277 | 142 | |
M28526.1 | - | 1 - 2 | 205 | 142 | |
M37780.1 | - | 1 - 2 | 189 | 142 |