The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013436.4 | NP_038464.1 | 22 - 23 | 3267 | 146 |
NM_205842.2 | NP_995314.1 | 23 - 24 | 3285 | 146 | |
XM_006712200.3 | XP_006712263.1 | 23 - 24 | 2771 | 146 | |
XM_006712201.3 | XP_006712264.1 | 22 - 23 | 2755 | 146 | |
GenBank mRNA | AB011159.1 | - | 23 - 24 | 3285 | 146 |
AB014509.1 | - | 22 - 23 | 2527 | 146 | |
AK001291.1 | - | 3 - 4 | 478 | 146 | |
AK002153.2 | - | 5 - 6 | 763 | 146 | |
AK292914.1 | - | 22 - 23 | 2757 | 146 | |
BC015025.1 | - | 22 - 23 | 2779 | 146 | |
BT007033.1 | - | 22 - 23 | 2508 | 146 |