The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005542.4 | NP_005533.2 | 3 - 4 | 746 | 93 |
XM_005249542.1 | XP_005249599.1 | 3 - 4 | 746 | 93 | |
XM_005249543.1 | XP_005249600.1 | 3 - 4 | 761 | 93 | |
XM_017012177.1 | XP_016867666.1 | 3 - 4 | 761 | 93 | |
GenBank mRNA | AK291675.1 | - | 3 - 4 | 618 | 93 |
AY112745.1 | - | 2 - 3 | 535 | 93 | |
BC001880.1 | - | 3 - 4 | 684 | 93 | |
BQ059075.1 | - | 2 - 3 | 279 | 93 | |
BT007227.1 | - | 2 - 3 | 535 | 93 |