Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IFNW1
Entrez Gene ID
3467
Gene Name
interferon omega 1
Gene Aliases
-
Chromosome Location
Chr.9: 21140632 - 21141901 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002177.2 NP_002168.1 1 - 1 219 107
GenBank mRNA A12140.1 - 1 - 1 463 107
AI860608.1 - 1 - 1 150 107

View Full Product Details