The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002233.3 | NP_002224.1 | 2 - 2 | 992 | 66 |
GenBank mRNA | AK313662.1 | - | 1 - 1 | 271 | 66 |
M55514.1 | - | 2 - 2 | 903 | 66 | |
M60450.1 | - | 1 - 1 | 48 | 66 |