The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004267.4 | NP_004258.2 | 1 - 2 | 763 | 101 |
GenBank mRNA | AB014679.2 | - | 1 - 2 | 72 | 101 |
AB014680.2 | - | 1 - 2 | 72 | 101 | |
AB021124.1 | - | 1 - 2 | 1208 | 101 | |
AK023268.1 | - | 1 - 2 | 713 | 101 | |
BC017499.1 | - | 1 - 2 | 444 | 101 | |
BC036930.1 | - | 1 - 2 | 674 | 101 | |
BC068604.1 | - | 1 - 2 | 713 | 101 |