Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNR2
Entrez Gene ID
1269
Gene Name
cannabinoid receptor 2
Gene Aliases
CB-2, CB2, CX5
Chromosome Location
Chr.1: 23870526 - 23913327 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001841.2 NP_001832.1 1 - 2 84 65
GenBank mRNA BC069722.1 - 1 - 2 26 65
X74328.1 - 1 - 2 83 65

View Full Product Details