Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHF20
Entrez Gene ID
51230
Gene Name
PHD finger protein 20
Gene Aliases
C20orf104, GLEA2, HCA58, NZF, TDRD20A, TZP
Chromosome Location
Chr.20: 35771999 - 35950377 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016436.4 NP_057520.2 14 - 15 2217 83
XM_005260421.2 XP_005260478.1 11 - 12 1827 83
XM_011528843.1 XP_011527145.1 13 - 14 1818 83
XM_011528844.1 XP_011527146.1 11 - 12 1889 83
XM_017027864.1 XP_016883353.1 15 - 16 2299 83
XM_017027865.1 XP_016883354.1 15 - 16 2500 83
XM_017027866.1 XP_016883355.1 14 - 15 2420 83
XM_017027867.1 XP_016883356.1 14 - 15 2255 83
XM_017027868.1 XP_016883357.1 12 - 13 2041 83
XM_017027869.1 XP_016883358.1 14 - 15 2214 83
XM_017027870.1 XP_016883359.1 14 - 15 2220 83
XM_017027871.1 XP_016883360.1 15 - 16 2300 83
GenBank mRNA AK097498.1 - 3 - 4 420 83
AK314503.1 - 14 - 15 2181 83
AY027523.1 - 13 - 14 2142 83
BC150178.1 - 14 - 15 2183 83

View Full Product Details