The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031210.5 | NP_112487.1 | 3 - 4 | 300 | 89 |
NR_052025.1 | 4 - 5 | 411 | 89 | ||
GenBank mRNA | AF253980.1 | - | 3 - 4 | 271 | 89 |
AF271779.1 | - | 3 - 4 | 295 | 89 | |
AJ272055.1 | - | 3 - 4 | 259 | 89 | |
AY860853.1 | - | 3 - 4 | 259 | 89 | |
BC017895.1 | - | 3 - 4 | 268 | 89 | |
BM685909.1 | - | 4 - 5 | 376 | 89 | |
BU944910.1 | - | 3 - 4 | 300 | 89 | |
BU960115.1 | - | 3 - 4 | 278 | 89 | |
EL951262.1 | - | 3 - 4 | 260 | 89 |