Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 1348006-1348517 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UVSSA
Entrez Gene ID
57654
Gene Name
UV stimulated scaffold protein A
Gene Aliases
KIAA1530, UVSS3
UniGene
Hs.380475
Gene Chromosome Location
Chr.4: 1345662-1388049 on Build GRCh38
Transcripts
NM_001317934.1 NM_001317935.1 NM_020894.3 XM_017008490.1 XM_017008491.1 XM_017008492.1 XM_017008493.1 XM_017008494.1 XM_017008496.1 XM_017008497.1 XM_017008498.1 XM_017008499.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1051244 c.17C>T p.S6L Chr.4: 1348108-1348108 on Build GRCh38
282199 c.89A>G p.K30R Chr.4: 1348180-1348180 on Build GRCh38
732571 c.67G>C p.E23Q Chr.4: 1348158-1348158 on Build GRCh38
6334384 c.98+5G>T p.? Chr.4: 1348194-1348194 on Build GRCh38
1212218 c.4G>T p.D2Y Chr.4: 1348095-1348095 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769316499 Chr.4: 1348063-1348063 on Build GRCh38
rs925068421 Chr.4: 1348463-1348463 on Build GRCh38
rs199574083 Chr.4: 1348146-1348146 on Build GRCh38
rs913276127 Chr.4: 1348445-1348445 on Build GRCh38
rs754033585 Chr.4: 1348088-1348088 on Build GRCh38
rs950831722 Chr.4: 1348172-1348172 on Build GRCh38
rs573032167 Chr.4: 1348036-1348036 on Build GRCh38
rs753074950 Chr.4: 1348075-1348078 on Build GRCh38
rs985035428 Chr.4: 1348032-1348032 on Build GRCh38
rs180993168 Chr.4: 1348492-1348492 on Build GRCh38
rs901113307 Chr.4: 1348222-1348222 on Build GRCh38
rs777126203 Chr.4: 1348239-1348239 on Build GRCh38
rs1001647212 Chr.4: 1348405-1348405 on Build GRCh38
rs764339228 Chr.4: 1348140-1348140 on Build GRCh38
rs550972367 Chr.4: 1348453-1348453 on Build GRCh38
rs762284452 Chr.4: 1348241-1348241 on Build GRCh38
rs531088325 Chr.4: 1348314-1348314 on Build GRCh38
rs960368034 Chr.4: 1348144-1348144 on Build GRCh38
rs1026086314 Chr.4: 1348149-1348149 on Build GRCh38
rs970435552 Chr.4: 1348404-1348408 on Build GRCh38
rs989066689 Chr.4: 1348288-1348288 on Build GRCh38
rs994057760 Chr.4: 1348223-1348223 on Build GRCh38
rs984721904 Chr.4: 1348234-1348234 on Build GRCh38
rs750475141 Chr.4: 1348164-1348164 on Build GRCh38
rs538778380 Chr.4: 1348442-1348442 on Build GRCh38
rs1033756212 Chr.4: 1348137-1348137 on Build GRCh38
rs371973794 Chr.4: 1348069-1348069 on Build GRCh38
rs769276364 Chr.4: 1348237-1348237 on Build GRCh38
rs779101623 Chr.4: 1348093-1348093 on Build GRCh38
rs373099973 Chr.4: 1348261-1348261 on Build GRCh38
rs747994631 Chr.4: 1348197-1348197 on Build GRCh38
rs778321304 Chr.4: 1348078-1348078 on Build GRCh38
rs189308280 Chr.4: 1348340-1348340 on Build GRCh38
rs954989184 Chr.4: 1348409-1348409 on Build GRCh38
rs770366519 Chr.4: 1348224-1348224 on Build GRCh38
rs888361586 Chr.4: 1348352-1348352 on Build GRCh38
rs933375007 Chr.4: 1348073-1348073 on Build GRCh38
rs910900056 Chr.4: 1348467-1348467 on Build GRCh38
rs749317573 Chr.4: 1348089-1348090 on Build GRCh38
rs1002496895 Chr.4: 1348300-1348300 on Build GRCh38
rs772263705 Chr.4: 1348046-1348046 on Build GRCh38
rs896128945 Chr.4: 1348318-1348318 on Build GRCh38
rs34635303 Chr.4: 1348343-1348344 on Build GRCh38
rs1053105266 Chr.4: 1348251-1348251 on Build GRCh38
rs543773523 Chr.4: 1348258-1348258 on Build GRCh38
rs757409533 Chr.4: 1348136-1348138 on Build GRCh38
rs1016835278 Chr.4: 1348361-1348361 on Build GRCh38
rs116804738 Chr.4: 1348147-1348147 on Build GRCh38
rs747439147 Chr.4: 1348235-1348235 on Build GRCh38
rs960246041 Chr.4: 1348418-1348418 on Build GRCh38
rs546886122 Chr.4: 1348364-1348364 on Build GRCh38
rs766301747 Chr.4: 1348183-1348183 on Build GRCh38
rs947072322 Chr.4: 1348312-1348312 on Build GRCh38
rs772694945 Chr.4: 1348118-1348118 on Build GRCh38
rs1043155291 Chr.4: 1348458-1348458 on Build GRCh38
rs569305186 Chr.4: 1348480-1348480 on Build GRCh38
rs779871040 Chr.4: 1348103-1348103 on Build GRCh38
rs1002165863 Chr.4: 1348135-1348135 on Build GRCh38
rs942336124 Chr.4: 1348475-1348476 on Build GRCh38
rs138607842 Chr.4: 1348108-1348108 on Build GRCh38
rs1026012546 Chr.4: 1348321-1348321 on Build GRCh38
rs367707267 Chr.4: 1348066-1348066 on Build GRCh38
rs780848645 Chr.4: 1348112-1348112 on Build GRCh38
rs570689863 Chr.4: 1348373-1348373 on Build GRCh38
rs753049966 Chr.4: 1348075-1348075 on Build GRCh38
rs138850155 Chr.4: 1348325-1348325 on Build GRCh38
rs1055019436 Chr.4: 1348489-1348489 on Build GRCh38
rs757606945 Chr.4: 1348089-1348089 on Build GRCh38
rs766362772 Chr.4: 1348289-1348289 on Build GRCh38
rs775564515 Chr.4: 1348060-1348060 on Build GRCh38
rs387907164 Chr.4: 1348185-1348185 on Build GRCh38
rs768295013 Chr.4: 1348059-1348059 on Build GRCh38
rs947518585 Chr.4: 1348265-1348265 on Build GRCh38
rs755830719 Chr.4: 1348401-1348401 on Build GRCh38
rs909466769 Chr.4: 1348038-1348038 on Build GRCh38
rs938827461 Chr.4: 1348266-1348266 on Build GRCh38
rs761048585 Chr.4: 1348139-1348139 on Build GRCh38
rs1043948875 Chr.4: 1348271-1348271 on Build GRCh38
rs768369135 Chr.4: 1348111-1348111 on Build GRCh38
rs777405757 Chr.4: 1348205-1348205 on Build GRCh38
rs747777373 Chr.4: 1348116-1348116 on Build GRCh38
rs371932049 Chr.4: 1348186-1348186 on Build GRCh38
rs763764498 Chr.4: 1348411-1348411 on Build GRCh38
rs377481343 Chr.4: 1348341-1348341 on Build GRCh38
rs1025523484 Chr.4: 1348351-1348351 on Build GRCh38
rs772561562 Chr.4: 1348129-1348129 on Build GRCh38
rs780844036 Chr.4: 1348196-1348196 on Build GRCh38
rs752268893 Chr.4: 1348374-1348374 on Build GRCh38
rs756171519 Chr.4: 1348306-1348306 on Build GRCh38
rs749484361 Chr.4: 1348334-1348335 on Build GRCh38
rs913523365 Chr.4: 1348440-1348440 on Build GRCh38
rs375317309 Chr.4: 1348071-1348071 on Build GRCh38
rs988973436 Chr.4: 1348424-1348424 on Build GRCh38
rs978450931 Chr.4: 1348446-1348446 on Build GRCh38
rs760954628 Chr.4: 1348078-1348078 on Build GRCh38
rs149266206 Chr.4: 1348119-1348119 on Build GRCh38
rs937877569 Chr.4: 1348484-1348484 on Build GRCh38
rs756548222 Chr.4: 1348077-1348078 on Build GRCh38
rs377711558 Chr.4: 1348230-1348230 on Build GRCh38
rs764340459 Chr.4: 1348086-1348086 on Build GRCh38
rs549301814 Chr.4: 1348315-1348315 on Build GRCh38
rs368110594 Chr.4: 1348117-1348117 on Build GRCh38
rs755705444 Chr.4: 1348201-1348201 on Build GRCh38
rs909260959 Chr.4: 1348034-1348034 on Build GRCh38
rs746834411 Chr.4: 1348051-1348051 on Build GRCh38
rs781695404 Chr.4: 1348131-1348131 on Build GRCh38
rs751474814 Chr.4: 1348184-1348184 on Build GRCh38
rs765616854 Chr.4: 1348163-1348163 on Build GRCh38
rs773103025 Chr.4: 1348246-1348246 on Build GRCh38
rs758964095 Chr.4: 1348319-1348319 on Build GRCh38
rs750921773 Chr.4: 1348247-1348247 on Build GRCh38
rs750601721 Chr.4: 1348095-1348095 on Build GRCh38
rs769684519 Chr.4: 1348173-1348173 on Build GRCh38
rs73793378 Chr.4: 1348211-1348211 on Build GRCh38
rs746066373 Chr.4: 1348216-1348219 on Build GRCh38
rs778975867 Chr.4: 1348178-1348178 on Build GRCh38
rs779890935 Chr.4: 1348047-1348047 on Build GRCh38
rs923538924 Chr.4: 1348383-1348383 on Build GRCh38

View Full Product Details