Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 1353336-1353839 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UVSSA
Entrez Gene ID
57654
Gene Name
UV stimulated scaffold protein A
Gene Aliases
KIAA1530, UVSS3
UniGene
Hs.380475
Gene Chromosome Location
Chr.4: 1345662-1388049 on Build GRCh38
Transcripts
NM_001317934.1 NM_001317935.1 NM_020894.3 XM_017008490.1 XM_017008491.1 XM_017008492.1 XM_017008493.1 XM_017008494.1 XM_017008495.1 XM_017008496.1 XM_017008497.1 XM_017008498.1 XM_017008499.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6300531 c.891G>A p.G297G Chr.4: 1353370-1353370 on Build GRCh38
6819376 c.919G>A p.V307M Chr.4: 1353398-1353398 on Build GRCh38
6099323 c.897C>G p.G299G Chr.4: 1353376-1353376 on Build GRCh38
6819379 c.912G>A p.T304T Chr.4: 1353391-1353391 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762609624 Chr.4: 1353394-1353395 on Build GRCh38
rs532916077 Chr.4: 1353676-1353676 on Build GRCh38
rs558079316 Chr.4: 1353492-1353492 on Build GRCh38
rs1028693699 Chr.4: 1353599-1353599 on Build GRCh38
rs150164998 Chr.4: 1353356-1353356 on Build GRCh38
rs977583913 Chr.4: 1353792-1353792 on Build GRCh38
rs971017241 Chr.4: 1353395-1353395 on Build GRCh38
rs1014783364 Chr.4: 1353571-1353571 on Build GRCh38
rs749042276 Chr.4: 1353378-1353378 on Build GRCh38
rs565718389 Chr.4: 1353377-1353377 on Build GRCh38
rs559247463 Chr.4: 1353425-1353425 on Build GRCh38
rs983334686 Chr.4: 1353398-1353398 on Build GRCh38
rs374676414 Chr.4: 1353379-1353379 on Build GRCh38
rs747642968 Chr.4: 1353368-1353368 on Build GRCh38
rs376465232 Chr.4: 1353442-1353442 on Build GRCh38
rs539989205 Chr.4: 1353468-1353468 on Build GRCh38
rs575310609 Chr.4: 1353819-1353819 on Build GRCh38
rs534930229 Chr.4: 1353542-1353542 on Build GRCh38
rs764317796 Chr.4: 1353433-1353433 on Build GRCh38
rs541902777 Chr.4: 1353702-1353702 on Build GRCh38
rs756047051 Chr.4: 1353366-1353366 on Build GRCh38
rs1033085096 Chr.4: 1353778-1353778 on Build GRCh38
rs752783215 Chr.4: 1353427-1353427 on Build GRCh38
rs551412333 Chr.4: 1353701-1353701 on Build GRCh38
rs556816908 Chr.4: 1353750-1353750 on Build GRCh38
rs369014709 Chr.4: 1353391-1353391 on Build GRCh38
rs771812101 Chr.4: 1353390-1353390 on Build GRCh38
rs766025783 Chr.4: 1353436-1353437 on Build GRCh38
rs887220785 Chr.4: 1353664-1353664 on Build GRCh38
rs879469949 Chr.4: 1353619-1353619 on Build GRCh38
rs936277744 Chr.4: 1353646-1353646 on Build GRCh38
rs987237418 Chr.4: 1353618-1353618 on Build GRCh38
rs953089672 Chr.4: 1353718-1353718 on Build GRCh38
rs372017016 Chr.4: 1353448-1353448 on Build GRCh38
rs553873246 Chr.4: 1353471-1353471 on Build GRCh38
rs753056861 Chr.4: 1353441-1353441 on Build GRCh38
rs921403138 Chr.4: 1353622-1353622 on Build GRCh38
rs773403730 Chr.4: 1353430-1353430 on Build GRCh38
rs1025189992 Chr.4: 1353534-1353534 on Build GRCh38
rs1002068916 Chr.4: 1353697-1353697 on Build GRCh38
rs6847321 Chr.4: 1353586-1353586 on Build GRCh38
rs779716014 Chr.4: 1353454-1353454 on Build GRCh38
rs753791967 Chr.4: 1353441-1353441 on Build GRCh38
rs372704748 Chr.4: 1353426-1353426 on Build GRCh38
rs778804926 Chr.4: 1353384-1353384 on Build GRCh38
rs561950822 Chr.4: 1353614-1353614 on Build GRCh38
rs749756010 Chr.4: 1353462-1353462 on Build GRCh38
rs752885294 Chr.4: 1353360-1353360 on Build GRCh38
rs571133677 Chr.4: 1353705-1353705 on Build GRCh38
rs868224685 Chr.4: 1353382-1353382 on Build GRCh38
rs189875131 Chr.4: 1353590-1353590 on Build GRCh38
rs756283314 Chr.4: 1353428-1353428 on Build GRCh38
rs757924417 Chr.4: 1353450-1353450 on Build GRCh38
rs556467504 Chr.4: 1353808-1353808 on Build GRCh38
rs774846460 Chr.4: 1353423-1353423 on Build GRCh38
rs950155543 Chr.4: 1353654-1353654 on Build GRCh38
rs1051743515 Chr.4: 1353628-1353628 on Build GRCh38
rs566067580 Chr.4: 1353455-1353455 on Build GRCh38
rs898231492 Chr.4: 1353678-1353678 on Build GRCh38
rs187433641 Chr.4: 1353521-1353521 on Build GRCh38
rs1009032852 Chr.4: 1353726-1353726 on Build GRCh38
rs926173849 Chr.4: 1353643-1353643 on Build GRCh38
rs538558435 Chr.4: 1353745-1353745 on Build GRCh38
rs991774302 Chr.4: 1353818-1353818 on Build GRCh38
rs999242422 Chr.4: 1353768-1353768 on Build GRCh38
rs73075573 Chr.4: 1353650-1353650 on Build GRCh38
rs894587298 Chr.4: 1353380-1353380 on Build GRCh38
rs369595773 Chr.4: 1353459-1353459 on Build GRCh38
rs955691666 Chr.4: 1353820-1353820 on Build GRCh38
rs766617413 Chr.4: 1353540-1353540 on Build GRCh38
rs574375586 Chr.4: 1353652-1353652 on Build GRCh38
rs373076268 Chr.4: 1353464-1353464 on Build GRCh38
rs1036194738 Chr.4: 1353680-1353680 on Build GRCh38
rs773734778 Chr.4: 1353401-1353401 on Build GRCh38
rs1021257642 Chr.4: 1353744-1353744 on Build GRCh38
rs763564095 Chr.4: 1353406-1353406 on Build GRCh38
rs769306986 Chr.4: 1353368-1353372 on Build GRCh38
rs898367415 Chr.4: 1353517-1353517 on Build GRCh38
rs992827173 Chr.4: 1353422-1353422 on Build GRCh38
rs1029034320 Chr.4: 1353717-1353717 on Build GRCh38
rs528971072 Chr.4: 1353627-1353627 on Build GRCh38
rs967744777 Chr.4: 1353588-1353588 on Build GRCh38
rs967582581 Chr.4: 1353621-1353621 on Build GRCh38
rs926143214 Chr.4: 1353798-1353798 on Build GRCh38
rs1003966412 Chr.4: 1353488-1353488 on Build GRCh38
rs766622274 Chr.4: 1353411-1353411 on Build GRCh38
rs772619822 Chr.4: 1353378-1353379 on Build GRCh38
rs770249281 Chr.4: 1353397-1353397 on Build GRCh38
rs745499970 Chr.4: 1353385-1353385 on Build GRCh38
rs776158676 Chr.4: 1353461-1353461 on Build GRCh38
rs1018682852 Chr.4: 1353746-1353746 on Build GRCh38
rs145597820 Chr.4: 1353359-1353359 on Build GRCh38
rs1033621320 Chr.4: 1353552-1353552 on Build GRCh38
rs1045829576 Chr.4: 1353662-1353662 on Build GRCh38
rs576763969 Chr.4: 1353566-1353566 on Build GRCh38
rs894548524 Chr.4: 1353479-1353479 on Build GRCh38
rs967142234 Chr.4: 1353763-1353763 on Build GRCh38

View Full Product Details