The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033341.4 | NP_203127.3 | 1 - 1 | 754 | 57 |
GenBank mRNA | AF420440.1 | - | 1 - 1 | 753 | 57 |
BC039318.1 | - | 1 - 1 | 524 | 57 | |
BC046168.1 | - | 1 - 1 | 498 | 57 | |
BC071665.1 | - | 1 - 1 | 302 | 57 |