The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001202504.1 | NP_001189433.1 | 3 - 4 | 272 | 64 |
NM_001318511.1 | NP_001305440.1 | 3 - 4 | 645 | 64 | |
NM_004715.4 | NP_004706.3 | 3 - 4 | 645 | 64 | |
NM_048368.3 | NP_430255.2 | 3 - 4 | 645 | 64 | |
XM_011526261.1 | XP_011524563.1 | 3 - 4 | 623 | 64 | |
GenBank mRNA | AF154115.1 | - | 3 - 4 | 581 | 64 |
BC032515.1 | - | 3 - 4 | 299 | 64 | |
BC047999.1 | - | 3 - 4 | 645 | 64 | |
BC052576.1 | - | 3 - 4 | 237 | 64 | |
BC063447.1 | - | 3 - 4 | 600 | 64 |