The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080652.3 | NP_542383.1 | 1 - 2 | 213 | 106 |
XM_017007437.1 | XP_016862926.1 | 1 - 2 | 213 | 106 | |
GenBank mRNA | AK172748.1 | - | 1 - 2 | 191 | 106 |
AK290878.1 | - | 1 - 2 | 191 | 106 | |
AK313954.1 | - | 1 - 2 | 106 | ||
AY358547.1 | - | 1 - 2 | 197 | 106 | |
BC008043.2 | - | 1 - 2 | 170 | 106 | |
BC019884.1 | - | 1 - 2 | 160 | 106 | |
BP274550.1 | - | 1 - 2 | 213 | 106 | |
BX647928.1 | - | 1 - 2 | 180 | 106 |