Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM41A
Entrez Gene ID
90407
Gene Name
transmembrane protein 41A
Gene Aliases
2900010K02Rik
Chromosome Location
Chr.3: 185482671 - 185499057 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080652.3 NP_542383.1 4 - 5 667 75
XM_017007437.1 XP_016862926.1 3 - 4 505 75
GenBank mRNA AK290878.1 - 4 - 5 645 75
AK313954.1 - 4 - 5 603 75
AL832782.1 - 1 - 2 126 75
AY358547.1 - 4 - 5 651 75
BC008043.2 - 4 - 5 624 75
BC019884.1 - 4 - 5 614 75
BX647128.1 - 2 - 3 317 75

View Full Product Details