The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017446.3 | NP_059142.2 | 4 - 5 | 532 | 125 |
NM_080794.3 | NP_542984.2 | 4 - 5 | 532 | 125 | |
XM_006724026.3 | XP_006724089.1 | 4 - 5 | 524 | 125 | |
XM_011529651.2 | XP_011527953.1 | 4 - 5 | 603 | 125 | |
GenBank mRNA | AF109357.1 | - | 4 - 5 | 506 | 125 |
AF239727.1 | - | 4 - 5 | 518 | 125 | |
AF270511.1 | - | 1 - 2 | 71 | 125 | |
AK000458.1 | - | 4 - 5 | 523 | 125 | |
AK225878.1 | - | 4 - 5 | 523 | 125 | |
BC004896.2 | - | 4 - 5 | 505 | 125 | |
BC107719.1 | - | 4 - 5 | 528 | 125 | |
BQ719968.1 | - | 4 - 5 | 560 | 125 | |
CD368044.1 | - | 6 - 7 | 125 |