The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002703.4 | NP_002694.3 | 8 - 9 | 1176 | 81 |
XM_011534387.1 | XP_011532689.1 | 8 - 9 | 953 | 81 | |
GenBank mRNA | AK222759.1 | - | 8 - 9 | 1115 | 81 |
AK290942.1 | - | 8 - 9 | 1031 | 81 | |
AK312297.1 | - | 8 - 9 | 1152 | 81 | |
BC004200.2 | - | 8 - 9 | 1048 | 81 | |
D13757.1 | - | 8 - 9 | 1138 | 81 | |
U00238.1 | - | 8 - 9 | 1067 | 81 |